This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04333980_m1
See other OBSCN GE Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04333980_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OBSCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098623.2 NP_001092093.2 54 - 55 14574 100
NM_001271223.2 NP_001258152.2 65 - 66 17445 100
NM_052843.3 NP_443075.3 54 - 55 14574 100
XM_005273287.4 XP_005273344.1 66 - 67 17825 100
XM_005273291.4 XP_005273348.1 66 - 67 17789 100
XM_005273298.4 XP_005273355.1 65 - 66 17516 100
XM_005273307.4 XP_005273364.1 61 - 62 16703 100
XM_006711822.3 XP_006711885.1 64 - 65 17249 100
XM_006711823.3 XP_006711886.1 64 - 65 17249 100
XM_006711827.3 XP_006711890.1 58 - 59 15664 100
XM_006711829.3 XP_006711892.1 57 - 58 15388 100
XM_011544297.2 XP_011542599.1 58 - 59 15665 100
XM_011544299.2 XP_011542601.1 57 - 58 15389 100
XM_017002443.1 XP_016857932.1 66 - 67 17870 100
XM_017002444.1 XP_016857933.1 66 - 67 17833 100
XM_017002445.1 XP_016857934.1 66 - 67 17870 100
XM_017002446.1 XP_016857935.1 66 - 67 17870 100
XM_017002447.1 XP_016857936.1 66 - 67 17867 100
XM_017002448.1 XP_016857937.1 66 - 67 17720 100
XM_017002449.1 XP_016857938.1 65 - 66 17607 100
XM_017002450.1 XP_016857939.1 65 - 66 17607 100
XM_017002451.1 XP_016857940.1 65 - 66 17607 100
XM_017002452.1 XP_016857941.1 65 - 66 17607 100
XM_017002453.1 XP_016857942.1 65 - 66 17594 100
XM_017002454.1 XP_016857943.1 65 - 66 17594 100
XM_017002455.1 XP_016857944.1 65 - 66 17594 100
XM_017002456.1 XP_016857945.1 65 - 66 17594 100
XM_017002457.1 XP_016857946.1 65 - 66 17594 100
XM_017002458.1 XP_016857947.1 64 - 65 17343 100
XM_017002459.1 XP_016857948.1 64 - 65 17343 100
XM_017002460.1 XP_016857949.1 64 - 65 17343 100
XM_017002461.1 XP_016857950.1 64 - 65 17330 100
XM_017002462.1 XP_016857951.1 64 - 65 17318 100
XM_017002463.1 XP_016857952.1 64 - 65 17297 100
XM_017002464.1 XP_016857953.1 64 - 65 17246 100
XM_017002465.1 XP_016857954.1 63 - 64 17079 100
XM_017002466.1 XP_016857955.1 63 - 64 17079 100
XM_017002467.1 XP_016857956.1 63 - 64 17079 100
XM_017002468.1 XP_016857957.1 62 - 63 16815 100
XM_017002469.1 XP_016857958.1 60 - 61 16287 100
XM_017002470.1 XP_016857959.1 55 - 56 14968 100
XM_017002471.1 XP_016857960.1 66 - 67 17869 100
XM_017002472.1 XP_016857961.1 66 - 67 17869 100
XM_017002473.1 XP_016857962.1 66 - 67 17869 100
GenBank mRNA AJ002535.1 - 55 - 56 14571 100

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100011861.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001888.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OBSCN

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01029446.hg.1 TC01001888.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01029446.hg.1 ENST00000284548.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029446.hg.1 ENST00000366707.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029446.hg.1 ENST00000366709.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029446.hg.1 ENST00000422127.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029446.hg.1 NM_001098623.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01029446.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000284548.15 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000284548.15 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187609.hg.1 TC0100011861.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0