This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04333988_g1
See other OBSCN GE Assays ›
Gene Symbol:
OBSCN
Gene Name:
obscurin, cytoskeletal calmodulin and titin-interacting RhoGEF
Gene Aliases:
ARHGEF30, UNC89
Chromosome Location:
Chr.1: 228208013 - 228381431 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04333988_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OBSCN
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001098623.2 NP_001092093.2 63 - 64 16547 88
NM_001271223.2 NP_001258152.2 74 - 75 19418 88
NM_052843.3 NP_443075.3 63 - 64 16547 88
XM_005273287.4 XP_005273344.1 75 - 76 19798 88
XM_005273291.4 XP_005273348.1 75 - 76 19762 88
XM_005273298.4 XP_005273355.1 74 - 75 19489 88
XM_005273307.4 XP_005273364.1 70 - 71 18676 88
XM_006711822.3 XP_006711885.1 73 - 74 19222 88
XM_006711823.3 XP_006711886.1 73 - 74 19222 88
XM_006711827.3 XP_006711890.1 67 - 68 17637 88
XM_006711829.3 XP_006711892.1 66 - 67 17361 88
XM_011544297.2 XP_011542599.1 67 - 68 17638 88
XM_011544299.2 XP_011542601.1 66 - 67 17362 88
XM_017002443.1 XP_016857932.1 75 - 76 19843 88
XM_017002444.1 XP_016857933.1 75 - 76 19806 88
XM_017002445.1 XP_016857934.1 75 - 76 19843 88
XM_017002446.1 XP_016857935.1 75 - 76 19843 88
XM_017002447.1 XP_016857936.1 75 - 76 19840 88
XM_017002448.1 XP_016857937.1 75 - 76 19693 88
XM_017002449.1 XP_016857938.1 74 - 75 19580 88
XM_017002450.1 XP_016857939.1 74 - 75 19580 88
XM_017002451.1 XP_016857940.1 74 - 75 19580 88
XM_017002452.1 XP_016857941.1 74 - 75 19580 88
XM_017002453.1 XP_016857942.1 74 - 75 19567 88
XM_017002454.1 XP_016857943.1 74 - 75 19567 88
XM_017002455.1 XP_016857944.1 74 - 75 19567 88
XM_017002456.1 XP_016857945.1 74 - 75 19567 88
XM_017002457.1 XP_016857946.1 74 - 75 19567 88
XM_017002458.1 XP_016857947.1 73 - 74 19316 88
XM_017002459.1 XP_016857948.1 73 - 74 19316 88
XM_017002460.1 XP_016857949.1 73 - 74 19316 88
XM_017002461.1 XP_016857950.1 73 - 74 19303 88
XM_017002462.1 XP_016857951.1 73 - 74 19291 88
XM_017002463.1 XP_016857952.1 73 - 74 19270 88
XM_017002464.1 XP_016857953.1 73 - 74 19219 88
XM_017002465.1 XP_016857954.1 72 - 73 19052 88
XM_017002466.1 XP_016857955.1 72 - 73 19052 88
XM_017002467.1 XP_016857956.1 72 - 73 19052 88
XM_017002468.1 XP_016857957.1 71 - 72 18788 88
XM_017002469.1 XP_016857958.1 69 - 70 18260 88
XM_017002470.1 XP_016857959.1 64 - 65 16941 88
XM_017002471.1 XP_016857960.1 75 - 76 19842 88
XM_017002472.1 XP_016857961.1 75 - 76 19842 88
XM_017002473.1 XP_016857962.1 75 - 76 19842 88
GenBank mRNA AB046859.2 - 3 - 4 319 88
AJ002535.1 - 64 - 65 16544 88
AL832357.1 - 8 - 9 963 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100011861.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01001888.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene OBSCN

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s) Process(es)

protein phosphorylation
multicellular organism development
regulation of Rho protein signal transduction
protein localization to M-band
positive regulation of apoptotic process
positive regulation of GTPase activity
sarcomere organization
regulation of small GTPase mediated signal transduction
mitophagy in response to mitochondrial depolarization
protein serine/threonine kinase activity
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
protein binding
calmodulin binding
ATP binding
structural constituent of muscle
ankyrin binding
titin binding
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01029457.hg.1 TC01001888.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01029457.hg.1 ENST00000284548.11 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029457.hg.1 ENST00000366707.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029457.hg.1 ENST00000366709.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029457.hg.1 ENST00000422127.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01029457.hg.1 NM_001098623.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR01029457.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000284548.15 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000284548.15 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000366707.8 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000422127.5 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 ENST00000570156.6 Ensembl
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 NM_001098623.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 NM_001271223.2 Refseq
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 NM_052843.3 Refseq
Affymetrix Human Clariom D Assay PSR0100187618.hg.1 TC0100011861.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0