This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04334669_m1
See other TRAPPC6A GE Assays ›
Gene Symbol:
TRAPPC6A
Gene Name:
trafficking protein particle complex 6A
Gene Aliases:
TRS33
Chromosome Location:
Chr.19: 45162928 - 45178302 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04334669_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAPPC6A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001270891.1 NP_001257820.1 3 - 4 291 112
NM_001270892.1 NP_001257821.1 2 - 3 265 112
NM_001270893.1 NP_001257822.1 2 - 3 223 112
NM_024108.2 NP_077013.1 3 - 4 333 112
GenBank mRNA BC001907.1 - 3 - 4 317 3534111 112
BC004450.1 - 3 - 4 318 2820830 112
BI462720.1 - 2 - 3 222 112
CN256686.1 - 2 - 3 260 112
CN256687.1 - 3 - 4 303 112
FJ418644.1 - 3 - 4 273 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900010926.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001615.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TRAPPC6A

Gene Ontology Categories:

Function(s) Process(es)

COPII vesicle coating
melanosome assembly
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19022039.hg.1 ENST00000006275.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19022039.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19022039.hg.1 TC19001615.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 TC1900010926.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay PSR1900190656.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000006275.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000585934.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000587818.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000588062.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 ENST00000592647.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270891.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270892.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_001270893.1 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq
Affymetrix Human Clariom D Assay TC1900010926.hg.1 NM_024108.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0