This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04394366_m1
See other LINC01336 GE Assays ›
Gene Symbol:
LINC01336
Gene Name:
long intergenic non-protein coding RNA 1336
Gene Aliases:
-
Chromosome Location:
Chr.5: 75047719 - 75052643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04394366_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01336
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_126375.1 1 - 2 214 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glucosaminyl (N-acetyl) transferase 4,
core 2
GCNT4 C2GNT3 -16.8kb Sense Chr. 5 - 75027464 - 75030899 on NCBI Build GRCh38
Downstream ankyrin repeat domain 31 ANKRD31 15.7kb Sense Chr. 5 - 75068297 - 75236897 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500011159.hg.1

Set Membership:

Amplicon length less than or equal to 70 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 NR_126375.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 NR_126375.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500011159.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay PSR0500177443.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500177443.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500177443.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay PSR0500177443.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500177443.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500177443.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay PSR0500177443.hg.1 TC0500011159.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500177439.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500177439.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay PSR0500177439.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500177439.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay PSR0500177439.hg.1 TC0500011159.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500177438.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500177438.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay PSR0500177438.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500177438.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay PSR0500177438.hg.1 TC0500011159.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500011159.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011159.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011159.hg.1 ENST00000503568.1 Ensembl
Affymetrix Human Clariom D Assay TC0500011159.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Clariom D Assay TC0500011159.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Clariom D Assay TC0500011159.hg.1 ENST00000506086.2 Ensembl
Affymetrix Human Clariom D Assay TC0500011159.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay TC0500011159.hg.1 NR_126375.1 Refseq
Affymetrix Human Clariom D Assay TC0500011159.hg.1 NR_126375.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0