This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04397260_m1
See other DERA GE Assays ›
Gene Symbol:
DERA
Gene Name:
deoxyribose-phosphate aldolase
Gene Aliases:
CGI-26, DEOC
Chromosome Location:
Chr.12: 15911172 - 16037381 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04397260_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DERA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300779.1 NP_001287708.1 6 - 7 824 96
NM_015954.3 NP_057038.2 7 - 8 953 96
GenBank mRNA AF132960.1 - 7 - 8 814 96
AK309182.1 - 7 - 8 869 96
BC041328.1 - 7 - 8 953 96
BC056234.1 - 7 - 8 795 96
BU928832.1 - 6 - 7 630 97

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DERA

Panther Classification:

Molecular Function -

aldolase

Gene Ontology Categories:

Function(s) Process(es)

pentose-phosphate shunt
deoxyribonucleotide catabolic process
carbohydrate catabolic process
deoxyribonucleoside catabolic process
deoxyribose phosphate catabolic process
deoxyribose-phosphate aldolase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2d9fq:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0