This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04397385_g1
See other BATF2 GE Assays ›
Gene Symbol:
BATF2
Gene Name:
basic leucine zipper ATF-like transcription factor 2
Gene Aliases:
SARI
Chromosome Location:
Chr.11: 64987945 - 64997045 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04397385_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BATF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300807.1 NP_001287736.1 1 - 2 207 83
GenBank mRNA AK300917.1 - 1 - 2 204 83

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100011197.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
defense response to protozoan
myeloid dendritic cell differentiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000301887.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000435842.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000435842.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000435842.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000435842.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000435842.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000435842.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527454.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527716.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527716.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527716.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527716.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527716.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000527716.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000534177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000534177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000534177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000534177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 ENST00000534177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300807.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300807.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300807.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300807.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300807.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300807.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300807.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300808.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300808.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300808.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300808.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300808.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_001300808.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100011197.hg.1 NM_138456.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-7gdsv:80/100.66.79.88:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0