This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04397714_m1
See other KMT5B GE Assays ›
Gene Symbol:
KMT5B
Gene Name:
lysine methyltransferase 5B
Gene Aliases:
CGI-85, CGI85, SUV420H1
Chromosome Location:
Chr.11: 68154863 - 68213904 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04397714_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: KMT5B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300907.1 NP_001287836.1 6 - 7 882 115
NM_001300908.1 NP_001287837.1 5 - 6 902 115
NM_001300909.1 NP_001287838.1 4 - 5 833 115
NM_016028.4 NP_057112.3 5 - 6 786 115
NM_017635.4 NP_060105.3 5 - 6 814 115
XM_005274035.3 XP_005274092.2 5 - 6 1034 115
XM_005274036.3 XP_005274093.2 4 - 5 965 115
XM_006718581.1 XP_006718644.1 4 - 5 696 115
XM_011545091.1 XP_011543393.1 4 - 5 608 115
XM_011545092.2 XP_011543394.1 4 - 5 798 115
XM_011545093.2 XP_011543395.1 5 - 6 1034 115
XM_011545094.2 XP_011543396.1 5 - 6 1034 115
XM_017017876.1 XP_016873365.1 6 - 7 1395 115
XM_017017877.1 XP_016873366.1 6 - 7 1181 115
XM_017017878.1 XP_016873367.1 6 - 7 1126 115
XM_017017879.1 XP_016873368.1 6 - 7 927 115
XM_017017880.1 XP_016873369.1 5 - 6 1034 115
GenBank mRNA AF264782.1 - 4 - 5 706 115
AK025183.1 - 4 - 5 474 115
AK299201.1 - 4 - 5 662 115
AK314336.1 - 5 - 6 729 115
BC002522.2 - 5 - 6 733 115
BC065287.1 - 5 - 6 738 115
BC087834.1 - 5 - 6 755 115
BC098121.1 - 4 - 5 535 40001705 115
BC099714.1 - 4 - 5 535 40001707 115
BC103498.1 - 4 - 5 535 40001706 115
BC104483.1 - 4 - 5 535 40001704 115

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the BC098121.1, BC104483.1, NM_001300907.1, NM_001300908.1, XM_011545093.2, XM_017017876.1, XM_017017877.1, XM_017017878.1, XM_017017879.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene KMT5B

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
muscle organ development
histone H4-K20 trimethylation
histone-lysine N-methyltransferase activity
histone methyltransferase activity (H4-K20 specific)

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0