This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04397990_g1
See other EIF3K GE Assays ›
Gene Symbol:
EIF3K
Gene Name:
eukaryotic translation initiation factor 3 subunit K
Gene Aliases:
ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001
Chromosome Location:
Chr.19: 38619072 - 38636959 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04397990_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300992.1 NP_001287921.1 1 - 2 252 75
NM_013234.3 NP_037366.1 1 - 2 252 75
GenBank mRNA AB019392.1 - 1 - 2 228 75
AF077051.1 - 1 - 2 228 75
AF085358.1 - 1 - 2 229 75
AF315506.1 - 1 - 2 60 75
AK026531.1 - 1 - 2 99 75
AK289554.1 - 1 - 2 130 75
AK301035.1 - 1 - 2 132 75
AK316344.1 - 1 - 2 137 75
AY245432.1 - 1 - 2 222 75
BC001031.2 - 1 - 2 116 3139375 75
BC007559.1 - 1 - 2 143 2988566 75
BF965565.1 - 1 - 2 81 75
BG470776.1 - 1 - 2 115 75
BG943109.1 - 1 - 2 253 75

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3K

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
translational initiation
regulation of translational initiation
translation initiation factor activity
protein binding
ribosome binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0