This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04397994_g1
See other EIF3K GE Assays ›
Gene Symbol:
EIF3K
Gene Name:
eukaryotic translation initiation factor 3 subunit K
Gene Aliases:
ARG134, EIF3-p28, EIF3S12, HSPC029, M9, MSTP001, PLAC-24, PLAC24, PRO1474, PTD001
Chromosome Location:
Chr.19: 38619072 - 38636959 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04397994_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3K
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001300992.1 NP_001287921.1 6 - 7 741 117
NM_001308393.1 NP_001295322.1 7 - 8 811 117
NM_013234.3 NP_037366.1 7 - 8 819 117

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900008019.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19000531.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3K

Panther Classification:

Molecular Function -

translation factor translation initiation factor translational protein

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
translational initiation
regulation of translational initiation
translation initiation factor activity
protein binding
ribosome binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19007196.hg.1 ENST00000248342.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19007196.hg.1 NM_013234.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19007196.hg.1 TC19000531.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000248342.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000588934.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000590134.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000592558.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000593149.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000614624.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 NM_001300992.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 NM_001308393.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 NM_013234.3 Refseq
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 TC1900008019.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000248342.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000588934.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000590134.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000592558.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000593149.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 ENST00000614624.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 NM_001300992.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 NM_001308393.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160268.hg.1 NM_013234.3 Refseq
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000248342.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000588934.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000592558.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000593149.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000614624.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 NM_001300992.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 NM_001308393.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 NM_013234.3 Refseq
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 TC1900008019.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000248342.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000588934.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000592558.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000593149.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 ENST00000614624.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 NM_001300992.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 NM_001308393.1 Refseq
Affymetrix Human Clariom D Assay PSR1900160270.hg.1 NM_013234.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0