This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04398450_m1
See other TMED3 GE Assays ›
Gene Symbol:
TMED3
Gene Name:
transmembrane p24 trafficking protein 3
Gene Aliases:
C15orf22, P24B, p24g4, p26
Chromosome Location:
Chr.15: 79311062 - 79419879 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04398450_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMED3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301203.1 NP_001288132.1 1 - 2 357 114
NM_007364.3 NP_031390.1 1 - 2 357 114
NR_125394.1 1 - 2 357 114
XM_017022040.1 XP_016877529.1 1 - 2 357 114
XM_017022041.1 XP_016877530.1 1 - 2 306 114
GenBank mRNA AJ132270.1 - 1 - 2 188 114
AK075529.1 - 1 - 2 260 114
AK222516.1 - 1 - 2 295 114
AK289434.1 - 1 - 2 276 114
AK297710.1 - 1 - 2 274 114
AK304148.1 - 1 - 2 263 114
AL109672.1 - 1 - 2 267 114
AY358974.1 - 1 - 2 267 114
BC000027.2 - 1 - 2 265 3507308 114
BC010853.1 - 1 - 2 302 3876465 114
BC017495.1 - 1 - 2 268 3921858 114
BC022232.1 - 1 - 2 292 3885305 114
BC111547.1 - 1 - 2 328 40005178 114
BM549126.1 - 1 - 2 194 114
BP260160.1 - 1 - 2 357 114
CR457384.1 - 1 - 2 169 114

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMED3

Panther Classification:

Molecular Function -

vesicle coat protein

Gene Ontology Categories:

Function(s)

ER to Golgi vesicle-mediated transport
retrograde vesicle-mediated transport, Golgi to ER
protein transport

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 ENST00000561560.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000299705.9 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000424155.6 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000536821.5 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000543455.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000558562.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 ENST00000561560.1 Ensembl
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_001301203.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NM_007364.3 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq
Affymetrix Human Clariom D Assay TC1500008003.hg.1 NR_125394.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0