This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04398644_m1
See other LRAT GE Assays ›
Gene Symbol:
LRAT
Gene Name:
lecithin retinol acyltransferase (phosphatidylcholine--retinol O-acyltransferase)
Gene Aliases:
LCA14
Chromosome Location:
Chr.4: 154740841 - 154753119 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04398644_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LRAT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301645.1 NP_001288574.1 2 - 3 599 113
NM_004744.4 NP_004735.2 2 - 3 793 113
GenBank mRNA AF071510.1 - 2 - 3 632 113
AK292598.1 - 2 - 3 793 113
AY546085.1 - 1 - 2 867 113
AY546086.1 - 2 - 3 764 113
BC031053.1 - 2 - 3 602 5272486 113

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene LRAT

Panther Classification:

Molecular Function -

acyltransferase

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
vitamin A metabolic process
visual perception
positive regulation of lipid transport
retinol metabolic process
retinoic acid metabolic process
response to stimulus
retinoic acid binding
O-palmitoyltransferase activity
transferase activity, transferring acyl groups
retinol binding
phosphatidylcholine-retinol O-acyltransferase activity
lecithin:11-cis retinol acyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-8rxhs:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0