This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04398746_m1
See other SS18L1 GE Assays ›
Gene Symbol:
SS18L1
Gene Name:
SS18L1, nBAF chromatin remodeling complex subunit
Gene Aliases:
CREST, LP2261
Chromosome Location:
Chr.20: 62143720 - 62182510 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04398746_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SS18L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301778.1 NP_001288707.1 10 - 11 1241 93
NM_198935.2 NP_945173.1 9 - 10 1136 93
NR_125980.1 10 - 11 1515 93
NR_125981.1 9 - 10 1136 93
XM_005260389.3 XP_005260446.1 9 - 10 1091 93
XM_005260391.1 XP_005260448.1 10 - 11 1275 93
XM_011528764.2 XP_011527066.1 10 - 11 2267 93
XM_011528767.1 XP_011527069.1 8 - 9 1077 93
XM_017027784.1 XP_016883273.1 9 - 10 1457 93
GenBank mRNA AB014593.1 - 9 - 10 1061 93
AK125656.1 - 10 - 11 1208 93
AK291485.1 - 9 - 10 1111 93
AK299881.1 - 9 - 10 1111 93
AY203931.1 - 6 - 7 823 93
BC034494.2 - 9 - 10 1090 4823298 93
BC068993.1 - 10 - 11 1453 5094932 93
BX640848.1 - 10 - 11 1273 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SS18L1

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
dendrite development
covalent chromatin modification
positive regulation of transcription, DNA-templated
positive regulation of dendrite morphogenesis
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline