This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04398772_m1
See other METTL6 GE Assays ›
Gene Symbol:
METTL6
Gene Name:
methyltransferase like 6
Gene Aliases:
-
Chromosome Location:
Chr.3: 15381275 - 15427580 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04398772_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: METTL6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301791.1 NP_001288720.1 3 - 4 623 120
NM_001301792.1 NP_001288721.1 3 - 4 651 120
NM_152396.3 NP_689609.2 3 - 4 623 120
XM_005264867.3 XP_005264924.1 3 - 4 623 120
XM_005264870.4 XP_005264927.1 3 - 4 623 120
XM_006712970.3 XP_006713033.1 3 - 4 499 120
XM_006712972.3 XP_006713035.1 3 - 4 623 120
XM_011533357.2 XP_011531659.1 3 - 4 623 120
XM_017005718.1 XP_016861207.1 3 - 4 499 120
XM_017005722.1 XP_016861211.1 3 - 4 623 120
XM_017005724.1 XP_016861213.1 3 - 4 623 120
GenBank mRNA AK057791.1 - 3 - 4 608 120
BC022400.1 - 3 - 4 580 4693314 120
BI913767.1 - 3 - 4 618 120
BU183521.1 - 3 - 4 561 120

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene METTL6

Panther Classification:

Molecular Function -

methyltransferase

Gene Ontology Categories:

Function(s) Process(es)

methylation
methyltransferase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-9hc65:80/100.66.78.230:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0