This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04399076_g1
See other FOSL1 GE Assays ›
Gene Symbol:
FOSL1
Gene Name:
FOS like 1, AP-1 transcription factor subunit
Gene Aliases:
FRA, FRA1, fra-1
Chromosome Location:
Chr.11: 65892136 - 65900526 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04399076_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FOSL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005438.4 NP_005429.1 2 - 3 486 91
GenBank mRNA AK222902.1 - 2 - 3 343 91
AK313718.1 - 2 - 3 379 91
BC016648.1 - 2 - 3 344 3925241 91
BQ937004.1 - 2 - 3 486 91
CR542257.1 - 2 - 3 299 91
CR542278.1 - 2 - 3 299 91
KU178407.1 - 2 - 3 299 91
X16707.1 - 2 - 3 333 91

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene FOSL1

Panther Classification:

Molecular Function -

basic leucine zipper transcription factor

Gene Ontology Categories:

Function(s) Process(es)

in utero embryonic development
regulation of transcription from RNA polymerase II promoter
transcription from RNA polymerase II promoter
chemotaxis
cellular defense response
vitellogenesis
female pregnancy
learning
positive regulation of cell proliferation
negative regulation of cell proliferation
response to mechanical stimulus
response to virus
response to gravity
cellular response to extracellular stimulus
response to progesterone
response to cytokine
response to drug
response to hydrogen peroxide
positive regulation of apoptotic process
positive regulation of cell cycle
positive regulation of transcription from RNA polymerase II promoter
positive regulation of sequence-specific DNA binding transcription factor activity
response to corticosterone
response to cAMP
placenta blood vessel development
pri-miRNA transcription from RNA polymerase II promoter
positive regulation of DNA-templated transcription, initiation
RNA polymerase II core promoter proximal region sequence-specific DNA binding
transcription factor activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcriptional activator activity, RNA polymerase II core promoter proximal region sequence-specific binding
transcription factor activity, sequence-specific DNA binding
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline