This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04399358_g1
See other GSTT1 GE Assays ›
Gene Symbol:
GSTT1
Gene Name:
glutathione S-transferase theta 1
Gene Aliases:
-
Chromosome Location:
Chr.22_KI270879v1: 270308 - 278486 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04399358_g1
Size
Availability Inventoried
Catalog # 4331182
Price 2,694.00
Your Price
Online offer:
2694.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: GSTT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_000853.3 NP_000844.2 4 - 5 609 71
NM_001293807.1 NP_001280736.1 4 - 5 567 71
NM_001293808.1 NP_001280737.1 3 - 4 460 71
NM_001293809.1 NP_001280738.1 2 - 3 370 71
NM_001293810.1 NP_001280739.1 3 - 4 521 71
NM_001293811.1 NP_001280740.1 3 - 4 458 71
NM_001293812.1 NP_001280741.1 4 - 5 548 71
GenBank mRNA AF435971.1 - 4 - 5 529 71
AJ867757.1 - 2 - 3 313 71
AJ874719.1 - 3 - 4 401 71
BC007065.1 - 4 - 5 569 3995846 71
BF343733.1 - 3 - 4 377 71
BG536639.1 - 4 - 5 501 71
BM765626.1 - 4 - 5 582 71
BX401239.2 - 4 - 5 496 71
CA313466.1 - 1 - 2 71
CD701199.1 - 2 - 3 355 71
CR456499.1 - 3 - 4 452 71
X79389.1 - 4 - 5 529 71

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC22000583.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene GSTT1

Gene Ontology Categories:

Function(s) Process(es)

glutathione metabolic process
cellular oxidant detoxification
glutathione derivative biosynthetic process
glutathione transferase activity
glutathione peroxidase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR22010281.hg.1 ENST00000248935.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010281.hg.1 ENST00000382792.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010281.hg.1 ENST00000439996.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010281.hg.1 ENST00000452369.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR22010281.hg.1 NM_000853.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR22010281.hg.1 TC22000583.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0