This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04399925_m1
See other SS18L1 GE Assays ›
Gene Symbol:
SS18L1
Gene Name:
SS18L1, nBAF chromatin remodeling complex subunit
Gene Aliases:
CREST, LP2261
Chromosome Location:
Chr.20: 62143720 - 62182510 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04399925_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SS18L1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_198935.2 NP_945173.1 4 - 5 475 93
NR_125980.1 5 - 6 854 93
NR_125981.1 4 - 5 475 93
XM_005260389.3 XP_005260446.1 4 - 5 430 93
XM_005260391.1 XP_005260448.1 5 - 6 614 93
XM_011528764.2 XP_011527066.1 5 - 6 1606 93
XM_011528767.1 XP_011527069.1 3 - 4 416 93
XM_017027784.1 XP_016883273.1 4 - 5 796 93
GenBank mRNA AB014593.1 - 4 - 5 400 93
AK291485.1 - 4 - 5 450 93
AK299881.1 - 4 - 5 450 93
AK308765.1 - 5 - 6 600 93
AY203931.1 - 1 - 2 162 93
BC034494.2 - 4 - 5 429 4823298 93
BC068993.1 - 5 - 6 792 5094932 93
DC375156.1 - 4 - 5 475 93

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SS18L1

Panther Classification:

Molecular Function -

transcription cofactor

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
dendrite development
covalent chromatin modification
positive regulation of transcription, DNA-templated
positive regulation of dendrite morphogenesis
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline