This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04400551_m1
See other MILR1 GE Assays ›
Gene Symbol:
MILR1
Gene Name:
mast cell immunoglobulin like receptor 1
Gene Aliases:
Allergin-1, C17orf60, MCA-32, MCA32
Chromosome Location:
Chr.17: 64449022 - 64474009 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04400551_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MILR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001085423.1 NP_001078892.1 5 - 6 810 112
NM_001291316.1 NP_001278245.1 4 - 5 525 112
NM_001291317.1 NP_001278246.1 4 - 5 540 112
XM_017024486.1 XP_016879975.1 5 - 6 903 112
XM_017024487.1 XP_016879976.1 5 - 6 823 112
GenBank mRNA AB542950.1 - 5 - 6 762 112
AB542951.1 - 4 - 5 471 112
AB542952.1 - 4 - 5 486 112
BC053534.1 - 4 - 5 704 112
BC130406.1 - 5 - 6 785 40146437 112
BC136594.1 - 5 - 6 785 9020584 112
BC144272.1 - 4 - 5 500 9052792 112
BE296352.1 - 2 - 3 311 112

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700008598.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin receptor superfamily

Gene Ontology Categories:

Function(s)

negative regulation of mast cell activation
mast cell degranulation

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000605096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000605096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000605096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000605096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000605096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000605096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000605096.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000612535.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000615220.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000616498.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000617071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000617071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000617071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000617071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000617071.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000619286.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000620304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000620304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000620304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000620304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000620304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 ENST00000620304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001085423.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291316.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008598.hg.1 NM_001291317.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0