This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04400652_m1
See other ELAVL4 GE Assays ›
Gene Symbol:
ELAVL4
Gene Name:
ELAV like neuron-specific RNA binding protein 4
Gene Aliases:
HUD, PNEM
Chromosome Location:
Chr.1: 50048014 - 50203786 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04400652_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ELAVL4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001294348.1 NP_001281277.1 1 - 2 342 85
NM_001324213.1 NP_001311142.1 1 - 2 342 85
NM_001324215.1 NP_001311144.1 1 - 2 342 85
NM_001324217.1 NP_001311146.1 1 - 2 342 85
GenBank mRNA AK298817.1 - 1 - 2 151 85
AK298902.1 - 1 - 2 342 85
AK309653.1 - 1 - 2 117 85

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AK298817.1, AK298902.1, NM_001294348.1, NM_001324213.1, NM_001324215.1, NM_001324217.1 transcripts.

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

translation initiation factor

Gene Ontology Categories:

Function(s) Process(es)

RNA processing
mRNA processing
nucleotide binding
RNA binding
mRNA 3'-UTR binding
AU-rich element binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4g6l8:80/100.66.76.118:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0