This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04400665_m1
See other RAD52 GE Assays ›
Gene Symbol:
RAD52
Gene Name:
RAD52 homolog, DNA repair protein
Gene Aliases:
-
Chromosome Location:
Chr.12: 911028 - 991195 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04400665_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAD52
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297419.1 NP_001284348.1 5 - 6 597 124
NM_001297420.1 NP_001284349.1 4 - 5 364 124
NM_001297421.1 NP_001284350.1 3 - 4 281 124
NM_001297422.1 NP_001284351.1 5 - 6 485 124
NM_134424.3 NP_602296.2 5 - 6 485 124
XM_005253720.4 XP_005253777.1 6 - 7 1359 124
XM_005253721.2 XP_005253778.1 5 - 6 585 124
XM_011520990.2 XP_011519292.1 6 - 7 621 124
XM_011520991.2 XP_011519293.1 5 - 6 487 124
XM_011520993.2 XP_011519295.1 5 - 6 436 124
XM_017019769.1 XP_016875258.1 6 - 7 742 124
XM_017019770.1 XP_016875259.1 5 - 6 436 124
XM_017019771.1 XP_016875260.1 5 - 6 436 124
GenBank mRNA AF125950.1 - 4 - 5 346 124
AK290047.1 - 5 - 6 405 124
AK299283.1 - 3 - 4 228 124
AY540753.1 - 4 - 5 347 124
BC104015.1 - 5 - 6 428 124
BC104016.1 - 5 - 6 506 124
BC104017.1 - 5 - 6 428 124
BC144458.1 - 3 - 4 534 124
L33262.1 - 5 - 6 615 124
U12134.1 - 4 - 5 378 124

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAD52

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA recombinase assembly
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
DNA double-strand break processing involved in repair via single-strand annealing
protein sumoylation
cellular response to oxidative stress
regulation of nucleotide-excision repair
DNA binding
protein binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0