This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04400670_m1
See other RAD52 GE Assays ›
Gene Symbol:
RAD52
Gene Name:
RAD52 homolog, DNA repair protein
Gene Aliases:
-
Chromosome Location:
Chr.12: 911028 - 991195 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04400670_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAD52
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297419.1 NP_001284348.1 6 - 7 717 115
NM_001297421.1 NP_001284350.1 4 - 5 401 115
NM_001297422.1 NP_001284351.1 6 - 7 605 115
NM_134424.3 NP_602296.2 6 - 7 605 115
XM_005253720.4 XP_005253777.1 7 - 8 1479 115
XM_005253721.2 XP_005253778.1 6 - 7 705 115
XM_011520990.2 XP_011519292.1 7 - 8 741 115
XM_011520991.2 XP_011519293.1 6 - 7 607 115
XM_011520993.2 XP_011519295.1 6 - 7 556 115
XM_011520995.2 XP_011519297.1 2 - 3 189 115
XM_017019769.1 XP_016875258.1 7 - 8 862 115
XM_017019770.1 XP_016875259.1 6 - 7 556 115
XM_017019771.1 XP_016875260.1 6 - 7 556 115
GenBank mRNA AF125948.1 - 5 - 6 728 115
AK290047.1 - 6 - 7 525 115
AK294709.1 - 6 - 7 550 115
AY540753.1 - 5 - 6 467 115
BC104015.1 - 6 - 7 548 115
BC104016.1 - 6 - 7 626 115
BC104017.1 - 6 - 7 548 115
L33262.1 - 6 - 7 735 115
U12134.1 - 5 - 6 498 115
U27516.1 - 5 - 6 497 115

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1200009566.hg.1
This assay targets the same location as HTA transcript cluster(s) TC12001087.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RAD52

Panther Classification:

Molecular Function -

DNA metabolism protein nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

DNA recombinase assembly
double-strand break repair
DNA recombination
cellular response to DNA damage stimulus
DNA double-strand break processing involved in repair via single-strand annealing
protein sumoylation
cellular response to oxidative stress
regulation of nucleotide-excision repair
DNA binding
protein binding
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12014370.hg.1 ENST00000358495.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12014370.hg.1 ENST00000430095.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12014370.hg.1 ENST00000536177.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12014370.hg.1 ENST00000539046.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12014370.hg.1 ENST00000545564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12014370.hg.1 NM_134424.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12014370.hg.1 TC12001087.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000228345.9 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000358495.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000430095.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000461568.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000463750.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000468231.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000481052.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000488642.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000536177.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000539046.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000542584.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000543912.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000545564.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297419.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297420.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297421.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297422.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_134424.3 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NR_123713.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000228345.9 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000358495.7 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000430095.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000461568.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000463750.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000468231.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000481052.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000488642.6 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000536177.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000539046.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000542584.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000543912.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 ENST00000545564.5 Ensembl
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297419.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297420.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297421.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_001297422.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NM_134424.3 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 NR_123713.1 Refseq
Affymetrix Human Clariom D Assay PSR1200171542.hg.1 TC1200009566.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0