This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04400973_m1
See other MAMSTR GE Assays ›
Gene Symbol:
MAMSTR
Gene Name:
MEF2 activating motif and SAP domain containing transcriptional regulator
Gene Aliases:
MASTR
Chromosome Location:
Chr.19: 48705718 - 48719898 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04400973_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAMSTR
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001130915.1 NP_001124387.1 9 - 10 1028 88
NM_001297753.1 NP_001284682.1 6 - 7 1141 88
NM_182574.2 NP_872380.1 7 - 8 1272 88
XM_011526807.2 XP_011525109.1 7 - 8 1277 88
XM_011526808.2 XP_011525110.1 9 - 10 1396 88
XM_011526809.2 XP_011525111.1 6 - 7 1075 88
XM_017026640.1 XP_016882129.1 8 - 9 1796 88
XM_017026641.1 XP_016882130.1 8 - 9 1757 88
XM_017026643.1 XP_016882132.1 8 - 9 1637 88
XM_017026644.1 XP_016882133.1 8 - 9 1607 88
XM_017026645.1 XP_016882134.1 5 - 6 1042 88
GenBank mRNA AK093389.1 - 7 - 8 1272 88
AK131427.1 - 9 - 10 1035 88
BC105056.1 - 5 - 6 696 8144059 88
BC105058.1 - 5 - 6 696 8144061 88
BC106047.1 - 6 - 7 1141 5786639 88
BC143443.1 - 4 - 5 501 9051951 88
BX280379.1 - 2 - 3 222 88

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MAMSTR

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
positive regulation of myotube differentiation
positive regulation of transcription from RNA polymerase II promoter
transcription factor activity, RNA polymerase II transcription factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0