This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04401205_s1
See other NUDT4 GE Assays ›
Gene Symbol:
NUDT4
Gene Name:
nudix hydrolase 4
Gene Aliases:
DIPP2, DIPP2alpha, DIPP2beta, HDCMB47P
Chromosome Location:
Chr.12: 93377923 - 93403248 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04401205_s1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NUDT4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001301022.1 NP_001287951.1 5 4266 108
NM_001301023.1 NP_001287952.1 5 4263 108
NM_001301024.1 NP_001287953.1 5 4166 108
NM_019094.5 NP_061967.3 5 4521 108
NM_199040.3 NP_950241.1 5 4524 108
GenBank mRNA AF191651.1 - 5 4109 108
AF191652.2 - 5 4110 108

Back To Top

More Information


Additional Information:

Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Both primers and probe map within a single exon

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NUDT4

Panther Classification:

Molecular Function -

phosphatase

Gene Ontology Categories:

Function(s) Process(es)

cyclic nucleotide metabolic process
calcium-mediated signaling
cyclic-nucleotide-mediated signaling
intracellular signal transduction
inositol phosphate metabolic process
regulation of RNA export from nucleus
intracellular transport
protein binding
diphosphoinositol-polyphosphate diphosphatase activity
snoRNA binding
metal ion binding
m7G(5')pppN diphosphatase activity
inositol diphosphate tetrakisphosphate diphosphatase activity
inositol bisdiphosphate tetrakisphosphate diphosphatase activity
inositol diphosphate pentakisphosphate diphosphatase activity
inositol-1-diphosphate-2,3,4,5,6-pentakisphosphate diphosphatase activity
inositol-3-diphosphate-1,2,4,5,6-pentakisphosphate diphosphatase activity
inositol-5-diphosphate-1,2,3,4,6-pentakisphosphate diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 1-diphosphatase activity
inositol-1,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity
inositol-3,5-bisdiphosphate-2,3,4,6-tetrakisphosphate 5-diphosphatase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0