This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04401476_m1
See other SLC25A17 GE Assays ›
Gene Symbol:
SLC25A17
Gene Name:
solute carrier family 25 member 17
Gene Aliases:
PMP34
Chromosome Location:
Chr.22: 40769630 - 40819399 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04401476_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC25A17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104235.1 6 - 7 797 74
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA269973.1 - 1 - 2 498 Chr. 22 - 40779062 - 40780255 [-] on Build GRCh38 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream melanin concentrating hormone receptor 1 MCHR1 GPR24,
MCH-1R,
MCH1R,
SLC-1,
SLC1
-96.2kb Anti-sense Chr. 22 - 40679178 - 40682814 on NCBI Build GRCh38
Downstream microRNA 4766 MIR4766 mir-4766 34.0kb Sense Chr. 22 - 40813883 - 40813958 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2200008796.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

fatty acid alpha-oxidation
translation
fatty acid beta-oxidation
ADP transport
ATP transport
fatty acid transport
coenzyme A transmembrane transport
FAD transmembrane transport
NAD transport
AMP transport
adenine nucleotide transmembrane transporter activity
structural constituent of ribosome
ATP transmembrane transporter activity
protein binding
ADP transmembrane transporter activity
coenzyme A transmembrane transporter activity
FAD transmembrane transporter activity
FMN transmembrane transporter activity
chaperone binding
NAD transporter activity
AMP transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 TC2200008796.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000263255.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000402844.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000412879.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000420970.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000426396.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000427084.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000430221.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000434193.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000435456.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000443810.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000447566.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000458600.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000491545.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000544408.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NM_001282726.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NM_001282727.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NM_006358.3 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104235.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104236.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104237.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104238.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000263255.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000402844.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000412879.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000420970.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000426396.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000427084.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000430221.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000434193.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000435456.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000443810.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000447566.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000458600.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000491545.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 ENST00000544408.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NM_001282726.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NM_001282727.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NM_006358.3 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104235.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104236.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104237.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163697.hg.1 NR_104238.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 TC2200008796.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000263255.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000402844.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000412879.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000420970.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000426396.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000427084.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000430221.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000435456.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000443810.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000447566.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000458600.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000491545.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000544408.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NM_001282726.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NM_001282727.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NM_006358.3 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104235.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104236.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104237.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104238.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000263255.10 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000402844.7 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000412879.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000420970.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000426396.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000427084.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000430221.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000435456.6 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000443810.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000447566.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000458600.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000491545.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 ENST00000544408.5 Ensembl
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NM_001282726.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NM_001282727.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NM_006358.3 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104235.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104236.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104237.1 Refseq
Affymetrix Human Clariom D Assay PSR2200163696.hg.1 NR_104238.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jbr9k:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0