This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04401723_m1
See other NEDD4 GE Assays ›
Gene Symbol:
NEDD4
Gene Name:
neural precursor cell expressed, developmentally down-regulated 4, E3 ubiquitin protein ligase
Gene Aliases:
NEDD4-1, RPF1
Chromosome Location:
Chr.15: 55826917 - 55993746 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04401723_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEDD4
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104302.1 17 - 18 3644 111
GenBank mRNA AL833293.1 - 18 - 19 2982 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream protogenin PRTG IGDCC5 -90.4kb Sense Chr. 15 - 55611540 - 55743649 on NCBI Build GRCh38
Downstream regulatory factor X7 RFX7 RFXDC2 252.4kb Sense Chr. 15 - 56090533 - 56247654 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1500009508.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
outflow tract morphogenesis
endocardial cushion development
protein monoubiquitination
protein targeting to lysosome
lysosomal transport
neuromuscular junction development
negative regulation of sodium ion transport
negative regulation of transcription from RNA polymerase II promoter in response to UV-induced DNA damage
positive regulation of phosphatidylinositol 3-kinase signaling
response to denervation involved in regulation of muscle adaptation
regulation of macroautophagy
protein ubiquitination
transmission of virus
negative regulation of vascular endothelial growth factor receptor signaling pathway
neuron projection development
receptor internalization
receptor catabolic process
cellular response to UV
regulation of ion transmembrane transport
T cell activation
regulation of membrane potential
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
glucocorticoid receptor signaling pathway
ubiquitin-dependent protein catabolic process via the multivesicular body sorting pathway
development involved in symbiotic interaction
positive regulation of protein catabolic process
positive regulation of nucleocytoplasmic transport
blood vessel morphogenesis
regulation of dendrite morphogenesis
regulation of synapse organization
progesterone receptor signaling pathway
response to calcium ion
protein K63-linked ubiquitination
regulation of potassium ion transmembrane transporter activity
negative regulation of sodium ion transmembrane transporter activity
protein binding
protein C-terminus binding
ligase activity
sodium channel inhibitor activity
protein domain specific binding
beta-2 adrenergic receptor binding
ubiquitin binding
phosphoserine binding
phosphothreonine binding
ubiquitin protein ligase activity
RNA polymerase binding
proline-rich region binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000338963.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000435532.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000503468.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000506154.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000508342.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000508871.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_001284338.1 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_001284339.1 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_001284340.1 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_006154.3 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_198400.3 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NR_104302.1 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 TC1500009508.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000338963.6 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000435532.7 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000503468.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000506154.1 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000508342.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 ENST00000508871.5 Ensembl
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_001284338.1 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_001284339.1 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_001284340.1 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_006154.3 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NM_198400.3 Refseq
Affymetrix Human Clariom D Assay PSR1500170177.hg.1 NR_104302.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline