This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04401906_m1
See other HACE1 GE Assays ›
Gene Symbol:
HACE1
Gene Name:
HECT domain and ankyrin repeat containing E3 ubiquitin protein ligase 1
Gene Aliases:
SPPRS
Chromosome Location:
Chr.6: 104682670 - 104859919 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04401906_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HACE1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_104424.1 16 - 17 2040 100
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA CN358104.1 - 3 - 4 416 Chr. 6 - 104777012 - 104784641 [-] on Build GRCh38 100

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream glutamate ionotropic receptor kainate type subunit 2 GRIK2 EAA4,
GLR6,
GLUK6,
GLUR6,
GluK2,
MRT6
-2.7mb Anti-sense Chr. 6 - 101393708 - 102070083 on NCBI Build GRCh38
Downstream LIN28B antisense RNA 1 LIN28B-AS1 C6orf220,
LINC00577,
dJ439I14.1
155.8kb Sense Chr. 6 - 104936294 - 104940527 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600012702.hg.1

Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

transcription, DNA-templated
regulation of transcription, DNA-templated
Golgi organization
cell cycle
protein ubiquitination
Rac protein signal transduction
regulation of cell migration
protein ubiquitination involved in ubiquitin-dependent protein catabolic process
membrane fusion
protein K48-linked ubiquitination
ubiquitin-protein transferase activity
protein binding
ligase activity
Rab GTPase binding
Rac GTPase binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000262903.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000369127.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000416605.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000517424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000517995.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000518503.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 NM_020771.3 Refseq
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 NR_104424.1 Refseq
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000262903.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000369127.8 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000416605.6 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000517424.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000517995.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 ENST00000518503.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 NM_020771.3 Refseq
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 NR_104424.1 Refseq
Affymetrix Human Clariom D Assay PSR0600187581.hg.1 TC0600012702.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline