This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04402448_m1
See other FBXL13 GE Assays ›
Gene Symbol:
FBXL13
Gene Name:
F-box and leucine rich repeat protein 13
Gene Aliases:
DRC6, Fbl13
Chromosome Location:
Chr.7: 102699226 - 103074841 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04402448_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: FBXL13
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_105043.1 3 - 4 341 114
XM_005250205.3 XP_005250262.1 3 - 4 333 114
XM_005250207.3 XP_005250264.1 3 - 4 333 114
XM_005250208.3 XP_005250265.1 3 - 4 333 114
XM_011515928.2 XP_011514230.1 3 - 4 333 114
XM_011515929.2 XP_011514231.1 3 - 4 333 114
XM_011515932.2 XP_011514234.1 3 - 4 333 114
XM_017011850.1 XP_016867339.1 3 - 4 277 114
XM_017011851.1 XP_016867340.1 3 - 4 333 114
XM_017011852.1 XP_016867341.1 3 - 4 483 114
GenBank mRNA BC026121.1 - 3 - 4 344 4826546 114
BG717734.1 - 3 - 4 346 114
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BG723564.1 - 3 - 4 318 Chr. 7 - 103025197 - 103074815 [-] on Build GRCh38 114

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nuclear factor,
erythroid 4
NFE4 NF-E4 -50.7kb Anti-sense Chr. 7 - 102973437 - 102978667 on NCBI Build GRCh38
Downstream armadillo repeat containing 10 ARMC10 PNAS-112,
PNAS112,
PSEC0198,
SVH
19.5kb Anti-sense Chr. 7 - 103074727 - 103099763 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline