This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04403190_m1
See other CTB-113P19.1 GE Assays ›
Gene Symbol:
CTB-113P19.1
Gene Name:
uncharacterized LOC101927096
Gene Aliases:
-
Chromosome Location:
Chr.5: 151676945 - 151687910 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04403190_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CTB-113P19.1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109873.1 1 - 2 104 102
GenBank mRNA BC039364.1 - 1 - 2 104 102
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB037618.1 - 1 - 2 95 Chr. 5 - 151676954 - 151686514 [+] on Build GRCh38 102
GenBankmRNA BI462126.1 - 1 - 2 109 Chr. 5 - 151676945 - 151686762 [+] on Build GRCh38 102

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream secreted protein acidic and cysteine rich SPARC BM-40,
OI17,
ON
Anti-sense Chr. 5 - 151661096 - 151687054 on NCBI Build GRCh38
Downstream antioxidant 1 copper chaperone ATOX1 ATX1,
HAH1
63.5kb Anti-sense Chr. 5 - 151742822 - 151758649 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500009128.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163840.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163840.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163840.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163840.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163840.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163840.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163840.hg.1 TC0500009128.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Clariom D Assay TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay TC0500009128.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163841.hg.1 TC0500009128.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 ENST00000510576.2 Ensembl
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 ENST00000518905.1 Ensembl
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 NR_109873.1 Refseq
Affymetrix Human Clariom D Assay PSR0500163842.hg.1 TC0500009128.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0