This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04403241_m1
See other MFSD2A GE Assays ›
Gene Symbol:
MFSD2A
Gene Name:
major facilitator superfamily domain containing 2A
Gene Aliases:
MCPH15, MFSD2, NLS1
Chromosome Location:
Chr.1: 39955112 - 39969968 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04403241_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MFSD2A
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109896.1 9 - 10 1186 83
GenBank mRNA AL834184.1 - 9 - 10 1140 83

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream v-myc avian myelocytomatosis viral oncogene lung carcinoma derived homolog MYCL L-Myc,
LMYC,
MYCL1,
bHLHe38
-65.1kb Anti-sense Chr. 1 - 39895424 - 39902015 on NCBI Build GRCh38
Downstream adenylate cyclase associated protein 1 CAP1 CAP,
CAP1-PEN
71.8kb Sense Chr. 1 - 40040070 - 40072649 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

fatty acid transport
phospholipid transport
transcytosis
transmembrane transport
establishment of blood-brain barrier
lipid transport across blood brain barrier
phospholipid transporter activity
symporter activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0