This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04403427_g1
See other ST7-AS2 GE Assays ›
Gene Symbol:
ST7-AS2
Gene Name:
ST7 antisense RNA 2
Gene Aliases:
ST7AS2, ST7OT2
Chromosome Location:
Chr.7: 117072072 - 117145592 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04403427_g1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ST7-AS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_109981.1 3 - 4 383 109

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream microRNA 6132 MIR6132 hsa-mir-6132 -97.6kb Anti-sense Chr. 7 - 117020211 - 117020319 on NCBI Build GRCh38
Downstream ST7 overlapping transcript 3 ST7-OT3 NCRNA00026,
ST7,
ST7OT3
64.2kb Anti-sense Chr. 7 - 117182681 - 117209937 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700012363.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700008885.hg.1 ENST00000622900.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay PSR0700189066.hg.1 TC0700012363.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700008885.hg.1 ENST00000622900.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000432541.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000433969.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000434993.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000442719.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000446784.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 ENST00000456577.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_002331.3 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109980.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq
Affymetrix Human Clariom D Assay TC0700012363.hg.1 NR_109981.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-nrcn4:80/100.66.78.247:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline