This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04403962_m1
See other HRAT17 GE Assays ›
Gene Symbol:
HRAT17
Gene Name:
heart tissue-associated transcript 17
Gene Aliases:
-
Chromosome Location:
Chr.7: 112954635 - 112995643 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04403962_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HRAT17
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110162.1 5 - 6 577 105
GenBank mRNA AA992588.1 - 1 - 2 105
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA HY205176.1 - 1 - 2 Chr. 7 - 112954648 - 112977203 [+] on Build GRCh38 105
GenBankmRNA HY205133.1 - 1 - 2 Chr. 7 - 112954648 - 112980088 [+] on Build GRCh38 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream base methyltransferase of 25S rRNA 2 homolog BMT2 C7orf60 -14.8kb Sense Chr. 7 - 112819147 - 112939877 on NCBI Build GRCh38
Downstream G protein-coupled receptor 85 GPR85 SREB,
SREB2
123.7kb Sense Chr. 7 - 113080413 - 113087778 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000447785.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 ENST00000451962.1 Ensembl
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq
Affymetrix Human Clariom D Assay TC0700012297.hg.1 NR_110162.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline