This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404036_m1
See other AADACL2-AS1 GE Assays ›
Gene Symbol:
AADACL2-AS1
Gene Name:
AADACL2 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 151751179 - 151928175 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404036_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AADACL2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110202.1 1 - 2 322 97
NR_110203.1 1 - 2 322 97
GenBank mRNA DB454604.1 - 1 - 2 322 97
HY022483.1 - 1 - 2 97
HY205836.1 - 2 - 3 97
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB454570.1 - 1 - 2 322 Chr. 3 - 151751474 - 151928175 [-] on Build GRCh38 97
GenBankmRNA HY029818.1 - 1 - 2 323 Chr. 3 - 151755066 - 151928175 [-] on Build GRCh38 97

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream arylacetamide deacetylase pseudogene 1 AADACP1 Anti-sense Chr. 3 - 151770456 - 151784894 on NCBI Build GRCh38
Downstream uncharacterized LOC101928166 LOC101928166 229.3kb Sense Chr. 3 - 152159464 - 152205429 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012825.hg.1

Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195968.hg.1 TC0300012825.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195973.hg.1 TC0300012825.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline