This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404037_m1
See other AADACL2-AS1 GE Assays ›
Gene Symbol:
AADACL2-AS1
Gene Name:
AADACL2 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 151751179 - 151928175 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404037_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AADACL2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110202.1 2 - 3 376 71
GenBank mRNA DB454604.1 - 2 - 3 376 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB454570.1 - 2 - 3 376 Chr. 3 - 151751474 - 151928175 [-] on Build GRCh38 71
GenBankmRNA BI460894.1 - 3 - 4 501 Chr. 3 - 151600852 - 151928168 [-] on Build GRCh38 71
GenBankmRNA HY029818.1 - 2 - 3 377 Chr. 3 - 151755066 - 151928175 [-] on Build GRCh38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream arylacetamide deacetylase pseudogene 1 AADACP1 Anti-sense Chr. 3 - 151770456 - 151784894 on NCBI Build GRCh38
Downstream arylacetamide deacetylase AADAC CES5A1,
DAC
39.4kb Anti-sense Chr. 3 - 151814008 - 151828488 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300012825.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195965.hg.1 TC0300012825.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300195964.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195964.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195964.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195964.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195964.hg.1 TC0300012825.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195967.hg.1 TC0300012825.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay PSR0300195966.hg.1 TC0300012825.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000475855.1 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 ENST00000483843.2 Ensembl
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110202.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq
Affymetrix Human Clariom D Assay TC0300012825.hg.1 NR_110203.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline