This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404201_m1
See other LAIR1 GE Assays ›
Gene Symbol:
LAIR1
Gene Name:
leukocyte associated immunoglobulin like receptor 1
Gene Aliases:
CD305, LAIR-1
Chromosome Location:
Chr.19: 54353624 - 54370556 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404201_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LAIR1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110280.2 4 - 5 970 75
GenBank mRNA BM918573.1 - 4 - 5 692 75

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream leukocyte immunoglobulin like receptor A4 LILRA4 CD85g,
ILT7
-17.8kb Sense Chr. 19 - 54333185 - 54339169 on NCBI Build GRCh38
Downstream tweety family member 1 TTYH1 55.4kb Anti-sense Chr. 19 - 54415431 - 54436719 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900011414.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

adaptive immune response
regulation of immune response
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000348231.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000391741.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000391742.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000391743.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000418556.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000420483.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000434277.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000436513.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000440716.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000460312.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000463489.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000467269.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000468656.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000474878.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000475389.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000498511.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289023.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289025.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289026.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289027.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_021706.4 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NR_110279.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NR_110280.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 TC1900011414.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000348231.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000391741.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000391742.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000391743.7 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000418556.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000420483.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000434277.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000436513.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000440716.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000460312.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000463489.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000467269.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000468656.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000474878.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000475389.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 ENST00000498511.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289023.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289025.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289026.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_001289027.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NM_021706.4 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NR_110279.2 Refseq
Affymetrix Human Clariom D Assay PSR1900196456.hg.1 NR_110280.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0