This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04404914_m1
See other LOC101928128 GE Assays ›
Gene Symbol:
LOC101928128
Gene Name:
uncharacterized LOC101928128
Gene Aliases:
-
Chromosome Location:
Chr.X: 85210706 - 85219289 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04404914_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101928128
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110651.1 3 - 4 493 119
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW082729.1 - 1 - 2 Chr. X - 85212048 - 85219289 [-] on Build GRCh38 119

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream spermidine/spermine N1-acetyl transferase-like 1 SATL1 -96.5kb Anti-sense Chr. X - 85092286 - 85116229 on NCBI Build GRCh38
Downstream zinc finger protein 711 ZNF711 CMPX1,
MRX97,
ZNF4,
ZNF5,
ZNF6,
Zfp711,
dJ75N13.1
24.5kb Sense Chr. X - 85243820 - 85273362 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00007780.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153157.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153157.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153157.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153157.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153157.hg.1 TC0X00007780.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay TC0X00007780.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153159.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153159.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153159.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153159.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153159.hg.1 TC0X00007780.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0X00153158.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153158.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153158.hg.1 ENST00000443247.1 Ensembl
Affymetrix Human Clariom D Assay PSR0X00153158.hg.1 NR_110651.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00153158.hg.1 TC0X00007780.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0