This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405037_m1
See other LOC101927239 GE Assays ›
Gene Symbol:
LOC101927239
Gene Name:
uncharacterized LOC101927239
Gene Aliases:
-
Chromosome Location:
Chr.17: 34169376 - 34183632 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405037_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101927239
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110748.1 4 - 5 731 76

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream acid sensing ion channel subunit 2 ASIC2 ACCN,
ACCN1,
ASIC2a,
BNC1,
BNaC1,
MDEG,
hBNaC1
-17.8kb Sense Chr. 17 - 33013087 - 34156806 on NCBI Build GRCh38
Downstream C-C motif chemokine ligand 2 CCL2 GDCF-2,
HC11,
HSMCR30,
MCAF,
MCP-1,
MCP1,
SCYA2,
SMC-CF
76.3kb Anti-sense Chr. 17 - 34255277 - 34257203 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700010371.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000579816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000579816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000579816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000580792.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000580792.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000580792.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 TC1700010371.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000580792.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181409.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 TC1700010371.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Clariom D Assay PSR1700181410.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000225823.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000359872.6 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000448983.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000566930.2 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000579816.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000579816.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000579816.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000580792.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000580792.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000583395.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 ENST00000584724.1 Ensembl
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_001094.4 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NM_183377.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NR_110748.1 Refseq
Affymetrix Human Clariom D Assay TC1700010371.hg.1 NR_110748.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-9dqhd:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline