This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405323_m1
See other RARB GE Assays ›
Gene Symbol:
RARB
Gene Name:
retinoic acid receptor beta
Gene Aliases:
HAP, MCOPS12, NR1B2, RARbeta1, RRB2
Chromosome Location:
Chr.3: 24829323 - 25597932 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405323_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RARB
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110892.1 3 - 4 924 60
GenBank mRNA CD014021.1 - 3 - 4 471 60

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC105376997 LOC105376997 -116.2kb Anti-sense Chr. 3 - 25383105 - 25384927 on NCBI Build GRCh38
Downstream topoisomerase (DNA) II beta TOP2B TOPIIB,
top2beta
17.2kb Anti-sense Chr. 3 - 25597905 - 25664936 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
ureteric bud development
glandular epithelial cell development
outflow tract septum morphogenesis
retinal pigment epithelium development
growth plate cartilage development
transcription initiation from RNA polymerase II promoter
signal transduction
positive regulation of cell proliferation
negative regulation of cell proliferation
striatum development
regulation of myelination
negative regulation of chondrocyte differentiation
embryonic hindlimb morphogenesis
multicellular organism growth
positive regulation of apoptotic process
negative regulation of apoptotic process
steroid hormone mediated signaling pathway
positive regulation of neuron differentiation
positive regulation of transcription from RNA polymerase II promoter
embryonic eye morphogenesis
retinoic acid receptor signaling pathway
embryonic digestive tract development
ventricular cardiac muscle cell differentiation
RNA polymerase II regulatory region sequence-specific DNA binding
DNA binding
steroid hormone receptor activity
retinoic acid receptor activity
drug binding
zinc ion binding
protein complex binding
retinoid X receptor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0