This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405332_g1
See other SLC14A2-AS1 GE Assays ›
Gene Symbol:
SLC14A2-AS1
Gene Name:
SLC14A2 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.18: 45438183 - 45507036 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405332_g1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC14A2-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_110899.1 4 - 5 342 88
GenBank mRNA AK126075.1 - 4 - 5 342 88
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB091714.1 - 4 - 5 342 Chr. 18 - 45483338 - 45507036 [-] on Build GRCh38 88

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream SET binding protein 1 SETBP1 MRD29,
SEB
-414.7kb Anti-sense Chr. 18 - 44680173 - 45068510 on NCBI Build GRCh38
Downstream solute carrier family 14 member 1 (Kidd blood group) SLC14A1 HUT11,
HsT1341,
JK,
RACH1,
RACH2,
UT-B1,
UT1,
UTE
239.6kb Anti-sense Chr. 18 - 45724123 - 45752520 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1800008574.hg.1

Set Membership:

Amplicon length between 86 and 100 Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000585897.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000585897.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000585897.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000588517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000588517.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000590257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000590257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000590257.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1800007194.hg.1 ENST00000590257.1 Ensembl
Affymetrix Human Clariom D Assay PSR1800158444.hg.1 NR_110899.2 Refseq
Affymetrix Human Clariom D Assay PSR1800158444.hg.1 TC1800008574.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800158444.hg.1 NR_110899.2 Refseq
Affymetrix Human Clariom D Assay PSR1800158445.hg.1 NR_110899.2 Refseq
Affymetrix Human Clariom D Assay PSR1800158445.hg.1 TC1800008574.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1800158445.hg.1 NR_110899.2 Refseq
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000585897.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000585897.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000585897.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000586386.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000588517.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000588517.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000590257.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000590257.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000590257.1 Ensembl
Affymetrix Human Clariom D Assay TC1800007194.hg.1 ENST00000590257.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-gm74s:80/100.66.78.154:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline