This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04405756_m1
See other LOC102723385 GE Assays ›
Gene Symbol:
LOC102723385
Gene Name:
uncharacterized LOC102723385
Gene Aliases:
-
Chromosome Location:
Chr.16: 19062754 - 19067691 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04405756_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102723385
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_119379.1 3 - 4 440 128
NR_119380.1 2 - 3 372 128
NR_119381.1 2 - 3 342 128
NR_119382.1 2 - 3 397 128
GenBank mRNA AW663894.1 - 1 - 2 128
FY212114.1 - 3 - 4 391 128
FY213069.1 - 2 - 3 323 128
FY213574.1 - 2 - 3 293 128
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AW103259.1 - 1 - 2 Chr. 16 - 19062754 - 19067357 [+] on Build GRCh38 128
GenBankmRNA AW573078.1 - 1 - 2 Chr. 16 - 19062748 - 19067376 [+] on Build GRCh38 128
GenBankmRNA FY213830.1 - 1 - 2 Chr. 16 - 19062862 - 19067641 [+] on Build GRCh38 128
GenBankmRNA AI651299.1 - 1 - 2 Chr. 16 - 19062758 - 19067691 [+] on Build GRCh38 128
GenBankmRNA FY211194.1 - Chr. - - [-] on Build 128
GenBankmRNA FY211806.1 - 1 - 2 Chr. 16 - 19062862 - 19067641 [+] on Build GRCh38 128
GenBankmRNA FY210902.1 - 1 - 2 Chr. 16 - 19062862 - 19067641 [+] on Build GRCh38 128
GenBankmRNA FY213705.1 - 3 - 4 392 Chr. 16 - 19062862 - 19067635 [-] on Build GRCh38 128
GenBankmRNA AA781211.1 - 1 - 2 Chr. 16 - 19062816 - 19067385 [+] on Build GRCh38 128

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transmembrane channel like 7 TMC7 Anti-sense Chr. 16 - 18983934 - 19063942 on NCBI Build GRCh38
Downstream coenzyme Q7,
hydroxylase
COQ7 CAT5,
CLK-1,
CLK1,
COQ10D8
3.6kb Anti-sense Chr. 16 - 19067595 - 19083097 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600009627.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173694.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173692.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173695.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173695.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173695.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173698.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay TC1600009627.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173693.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000568971.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 ENST00000571934.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173699.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173696.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173696.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173696.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173696.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173696.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173696.hg.1 ENST00000576433.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173696.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 ENST00000567047.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 NR_119379.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 NR_119380.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173700.hg.1 NR_119381.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173697.hg.1 NR_119382.1 Refseq
Affymetrix Human Clariom D Assay PSR1600173697.hg.1 TC1600009627.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1600173697.hg.1 NR_119382.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline