This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406091_m1
See other LOC100507065 GE Assays ›
Gene Symbol:
LOC100507065
Gene Name:
uncharacterized LOC100507065
Gene Aliases:
-
Chromosome Location:
Chr.12: 65466817 - 65642372 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406091_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC100507065
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120433.1 4 - 5 568 98
GenBank mRNA BU619406.1 - 1 - 2 98

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream methionine sulfoxide reductase B3 MSRB3 DFNB74 -32.7kb Anti-sense Chr. 12 - 65278643 - 65466907 on NCBI Build GRCh38
Downstream uncharacterized LOC105369187 LOC105369187 5.5kb Sense Chr. 12 - 65544238 - 65557311 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-b97mm:80/100.66.72.101:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0