This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406369_m1
See other LOC102724301 GE Assays ›
Gene Symbol:
LOC102724301
Gene Name:
uncharacterized LOC102724301
Gene Aliases:
-
Chromosome Location:
Chr.11: 119729583 - 119739623 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406369_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC102724301
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120587.1 1 - 2 229 121
GenBank mRNA BI194967.1 - 1 - 2 228 121
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA BM874533.1 - 1 - 2 217 Chr. 11 - 119729595 - 119739552 [+] on Build GRCh38 121

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nectin cell adhesion molecule 1 NECTIN1 CD111,
CLPED1,
ED4,
HIgR,
HV1S,
HVEC,
OFC7,
PRR,
PRR1,
PVRL1,
PVRR,
PVRR1,
SK-12,
nectin-1
-858.0 Anti-sense Chr. 11 - 119638098 - 119728725 on NCBI Build GRCh38
Downstream tripartite motif containing 29 TRIM29 ATDC 377.5kb Anti-sense Chr. 11 - 120111286 - 120138179 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100009262.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169429.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100169429.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169429.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100169429.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169429.hg.1 TC1100009262.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100169434.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100169434.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169434.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100169434.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169434.hg.1 TC1100009262.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009262.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay TC1100009262.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169431.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100169431.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169431.hg.1 ENST00000533253.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100169431.hg.1 NR_120587.1 Refseq
Affymetrix Human Clariom D Assay PSR1100169431.hg.1 TC1100009262.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4x8c5:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0