This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406396_m1
See other LINCR-0001 GE Assays ›
Gene Symbol:
LINCR-0001
Gene Name:
uncharacterized LINCR-0001
Gene Aliases:
-
Chromosome Location:
Chr.8: 10474565 - 10481974 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406396_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINCR-0001
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120604.1 1 - 2 63 103
GenBank mRNA DA212384.1 - 1 - 2 63 103

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream methionine sulfoxide reductase A MSRA PMSR -45.7kb Sense Chr. 8 - 10054224 - 10428891 on NCBI Build GRCh38
Downstream protease,
serine 55
PRSS55 CT153,
T-SP1,
TSP1,
UNQ9391
48.2kb Sense Chr. 8 - 10525331 - 10554166 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800012262.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000518098.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000518098.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000518098.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184538.hg.1 TC0800012262.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800184536.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184536.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184536.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184536.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184536.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184536.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184536.hg.1 TC0800012262.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000517732.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000518098.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000518098.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000518098.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay TC0800012262.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184537.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184537.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184537.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184537.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184537.hg.1 ENST00000524047.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800184537.hg.1 NR_120604.1 Refseq
Affymetrix Human Clariom D Assay PSR0800184537.hg.1 TC0800012262.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-dd86746ff-77ss8:80/100.66.75.30:80.
git-commit: 04e44c58f20fafe66519586ffe6db5d8a90e52ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.53.1-Offline