This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406549_m1
See other NEURL1-AS1 GE Assays ›
Gene Symbol:
NEURL1-AS1
Gene Name:
NEURL1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 103479603 - 103517442 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406549_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer:
434.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEURL1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120675.1 1 - 2 59 65
NR_120676.1 1 - 2 59 65
GenBank mRNA BX089018.1 - 1 - 2 59 65

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream calcium homeostasis modulator 3 CALHM3 FAM26A,
bA225H22.7
-36.1kb Sense Chr. 10 - 103472804 - 103479240 on NCBI Build GRCh38
Downstream SH3 and PX domains 2A SH3PXD2A FISH,
SH3MD1,
TKS5
76.6kb Sense Chr. 10 - 103594027 - 103855464 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000011721.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180252.hg.1 TC1000011721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000180251.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180251.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180251.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000180251.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180251.hg.1 TC1000011721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000180254.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180254.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180254.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180254.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay PSR1000180254.hg.1 TC1000011721.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120676.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline