| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000447820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000447820.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120676.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120676.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1000011721.hg.1 |
NR_120676.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000447820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000447820.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120676.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120676.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1000011721.hg.1 |
NR_120676.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180249.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180249.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180249.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180249.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180249.hg.1 |
ENST00000453753.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1000180249.hg.1 |
NR_120675.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1000180249.hg.1 |
TC1000011721.hg.1 |
Clariom D TC |