This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406552_m1
See other NEURL1-AS1 GE Assays ›
Gene Symbol:
NEURL1-AS1
Gene Name:
NEURL1 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.10: 103479603 - 103517442 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406552_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEURL1-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_120676.1 2 - 3 121 102
GenBank mRNA BX095052.1 - 2 - 3 95 102

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000447820.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 ENST00000453753.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120675.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120676.1 Refseq
Affymetrix Human Clariom D Assay TC1000011721.hg.1 NR_120676.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline