This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406679_m1
See other PTPRD-AS1 GE Assays ›
Gene Symbol:
PTPRD-AS1
Gene Name:
PTPRD antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.9: 8858018 - 8861727 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406679_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PTPRD-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_121600.1 1 - 2 309 108
GenBank mRNA DB495850.1 - 1 - 2 185 108
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA118661.1 - 1 - 2 265 Chr. 9 - 8858062 - 8860983 [+] on Build GRCh38 108
GenBankmRNA BF381765.1 - 178 Chr. - - [-] on Build 108
GenBankmRNA DB472955.1 - 1 - 2 178 Chr. 9 - 8858149 - 8861062 [+] on Build GRCh38 108

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream transmembrane protein 261 TMEM261 C9orf123 -1.1mb Anti-sense Chr. 9 - 7796490 - 7799806 on NCBI Build GRCh38
Downstream uncharacterized LOC105375972 LOC105375972 937.7kb Sense Chr. 9 - 9799392 - 9803791 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-gwzxr:80/100.66.76.73:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0