This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406808_m1
See other PACRG-AS3 GE Assays ›
Gene Symbol:
PACRG-AS3
Gene Name:
PACRG antisense RNA 3
Gene Aliases:
-
Chromosome Location:
Chr.6: 163181645 - 163192002 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406808_m1
Size
Availability Made To Order
Catalog # 4426961
Price 434.00
Your Price
Online offer: 407.65
407.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PACRG-AS3
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_122120.1 1 - 2 122 69
GenBank mRNA AK311212.1 - 1 - 2 122 69
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DA278000.1 - 1 - 2 122 Chr. 6 - 163183293 - 163192002 [-] on Build GRCh38 69
GenBankmRNA DB480283.1 - 1 - 2 65 Chr. 6 - 163166054 - 163191943 [-] on Build GRCh38 69

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream PACRG antisense RNA 2 PACRG-AS2 -136.7kb Sense Chr. 6 - 163042980 - 163054161 on NCBI Build GRCh38
Downstream PACRG antisense RNA 1 PACRG-AS1 118.0kb Sense Chr. 6 - 163309985 - 163324473 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600013814.hg.1

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000622071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000622071.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195319.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195319.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195319.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195319.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195319.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195319.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195319.hg.1 TC0600013814.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000622071.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000622071.1 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay TC0600013814.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195325.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195325.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195325.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195325.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195325.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195325.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195325.hg.1 TC0600013814.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600195327.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195327.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195327.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195327.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195327.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195327.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195327.hg.1 TC0600013814.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000419182.5 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000612841.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000614038.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000620150.1 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 ENST00000622490.4 Ensembl
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 NR_122120.1 Refseq
Affymetrix Human Clariom D Assay PSR0600195322.hg.1 TC0600013814.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-st8h5:80/100.66.79.144:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0