This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04406942_m1
See other LINC01609 GE Assays ›
Gene Symbol:
LINC01609
Gene Name:
long intergenic non-protein coding RNA 1609
Gene Aliases:
-
Chromosome Location:
Chr.8: 111098961 - 111236203 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04406942_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC01609
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125418.1 1 - 2 114 105
GenBank mRNA DB448904.1 - 1 - 2 82 105

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream long intergenic non-protein coding RNA 1608 LINC01608 -89.1kb Sense Chr. 8 - 110937690 - 111027433 on NCBI Build GRCh38
Downstream CUB and Sushi multiple domains 3 CSMD3 986.7kb Sense Chr. 8 - 112222928 - 113437499 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0800011500.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178241.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178241.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178241.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178241.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178241.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178241.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178241.hg.1 TC0800011500.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Clariom D Assay TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay TC0800011500.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178242.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178242.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178242.hg.1 ENST00000522778.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178242.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178242.hg.1 TC0800011500.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0800178239.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178239.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178239.hg.1 ENST00000519506.1 Ensembl
Affymetrix Human Clariom D Assay PSR0800178239.hg.1 NR_125418.1 Refseq
Affymetrix Human Clariom D Assay PSR0800178239.hg.1 TC0800011500.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-7669c5b596-nbv7w:80/100.66.76.242:80.
git-commit: 17e8f6c92ed407c129a469238c057b56fdf8abbb
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.36.2-2025.09.39-1.0