This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407138_m1
See other LOC101926915 GE Assays ›
Gene Symbol:
LOC101926915
Gene Name:
uncharacterized LOC101926915
Gene Aliases:
-
Chromosome Location:
Chr.6: 46276324 - 46365508 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407138_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LOC101926915
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_125838.1 3 - 4 571 74
GenBank mRNA BC043004.2 - 3 - 4 571 74

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream ectonucleotide pyrophosphatase/phosphodiesterase 5 (putative) ENPP5 NPP-5,
NPP5
-190.1kb Anti-sense Chr. 6 - 46159182 - 46171045 on NCBI Build GRCh38
Downstream uncharacterized LOC101926898 LOC101926898 126.5kb Sense Chr. 6 - 46492052 - 46533486 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0600008161.hg.1

Set Membership:

Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR0600157107.hg.1 NR_125838.1 Refseq
Affymetrix Human Clariom D Assay PSR0600157107.hg.1 NR_125838.1 Refseq
Affymetrix Human Clariom D Assay PSR0600157107.hg.1 TC0600008161.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-wt24k:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline