This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407738_m1
See other HDAC11-AS1 GE Assays ›
Gene Symbol:
HDAC11-AS1
Gene Name:
HDAC11 antisense RNA 1
Gene Aliases:
-
Chromosome Location:
Chr.3: 13476987 - 13480053 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407738_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HDAC11-AS1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046690.1 1 - 2 412 111
GenBank mRNA BX104155.1 - 1 - 2 412 111
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AI024915.1 - 1 - 2 Chr. 3 - 13476985 - 13479955 [+] on Build GRCh38 111
GenBankmRNA AI418468.1 - 1 - 2 Chr. 3 - 13476982 - 13479965 [+] on Build GRCh38 111

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream nucleoporin 210 NUP210 GP210,
POM210
-56.7kb Sense Chr. 3 - 13316230 - 13420319 on NCBI Build GRCh38
Downstream histone deacetylase 11 HDAC11 HD11 118.0 Anti-sense Chr. 3 - 13480171 - 13506424 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300010341.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0300010341.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010341.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0300010341.hg.1 NR_046690.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0300010341.hg.1 NR_046690.1 Refseq
Affymetrix Human Clariom D Assay TC0300010341.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010341.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Clariom D Assay TC0300010341.hg.1 NR_046690.1 Refseq
Affymetrix Human Clariom D Assay TC0300010341.hg.1 NR_046690.1 Refseq
Affymetrix Human Clariom D Assay PSR0300175392.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300175392.hg.1 NR_046690.1 Refseq
Affymetrix Human Clariom D Assay PSR0300175392.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300175392.hg.1 NR_046690.1 Refseq
Affymetrix Human Clariom D Assay PSR0300175392.hg.1 TC0300010341.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300175393.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300175393.hg.1 NR_046690.1 Refseq
Affymetrix Human Clariom D Assay PSR0300175393.hg.1 ENST00000424112.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300175393.hg.1 NR_046690.1 Refseq
Affymetrix Human Clariom D Assay PSR0300175393.hg.1 TC0300010341.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-49tpj:80/100.66.77.222:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline