This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs04407764_m1
See other HAO2-IT1 GE Assays ›
Gene Symbol:
HAO2-IT1
Gene Name:
HAO2 intronic transcript 1
Gene Aliases:
-
Chromosome Location:
Chr.1: 119368946 - 119370331 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs04407764_m1
Size
Availability Made To Order
Catalog # 4426961
Price 4,565.00
Your Price
Online offer:
4565.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HAO2-IT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_046780.1 1 - 2 61 123
GenBank mRNA AW300409.1 - 1 - 2 123

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream uncharacterized LOC101929147 LOC101929147 -93.0kb Sense Chr. 1 - 119140396 - 119275973 on NCBI Build GRCh38
Downstream hydroxy-delta-5-steroid dehydrogenase,
3 beta- and steroid delta-isomerase 2
HSD3B2 HSD3B,
HSDB,
SDR11E2
44.6kb Sense Chr. 1 - 119414931 - 119423039 on NCBI Build GRCh38

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100009609.hg.1

Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0100009609.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009609.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100009609.hg.1 NR_046780.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100009609.hg.1 NR_046780.1 Refseq
Affymetrix Human Clariom D Assay TC0100009609.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009609.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Clariom D Assay TC0100009609.hg.1 NR_046780.1 Refseq
Affymetrix Human Clariom D Assay TC0100009609.hg.1 NR_046780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100169154.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169154.hg.1 NR_046780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100169154.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169154.hg.1 NR_046780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100169154.hg.1 TC0100009609.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0100169155.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169155.hg.1 NR_046780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100169155.hg.1 ENST00000419144.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100169155.hg.1 NR_046780.1 Refseq
Affymetrix Human Clariom D Assay PSR0100169155.hg.1 TC0100009609.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0